missing translation for 'onlineSavingsMsg'
Learn More

Benzamide oxime, 97%, Thermo Scientific Chemicals

Catalog Number p-7077413
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
613-92-3
C7H8N2O
136.15
MFCD00031485 MFCD00474011
MXOQNVMDKHLYCZ-UHFFFAOYSA-N
benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
7259353
CHEBI:83354
N'-hydroxybenzenecarboximidamide
N\C(=N/O)C1=CC=CC=C1
This item is not returnable. View return policy
613-92-3
C7H8N2O
136.15
MFCD00031485 MFCD00474011
MXOQNVMDKHLYCZ-UHFFFAOYSA-N
benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
7259353
CHEBI:83354
N'-hydroxybenzenecarboximidamide
N\C(=N/O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 74.0°C to 77.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Linear Formula C6H5C(NH2)=NOH
Quantity 1 g
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material Benzamide oxime
compliance-icons
Product Identifier
  • Benzamide oxime, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'