missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride Hydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-3735547
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
206752-36-5, 1670-14-0
C7H9ClN2
156.61
MFCD00066285
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
16219042
benzenecarboximidamide;hydrate;hydrochloride
Cl.NC(=N)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
206752-36-5, 1670-14-0
C7H9ClN2
156.61
MFCD00066285
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
16219042
benzenecarboximidamide;hydrate;hydrochloride
Cl.NC(=N)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 86°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (on dry substance) (Argentometry)
Packaging Glass Bottle
Linear Formula HCl·xH2O
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 156.61
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzamidine hydrochloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'