missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[a]pyrene-d12, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Benzo(a)pyrene D12 |
| CAS | 63466-71-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 2H12 |
| Quantity | 10 mg |
| Synonym | Benzo[a]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Benzo[a]pyrene-d12 (9CI), Benzo[a]pyrene d12, Perdeuterated benzo[a]pyrene, [12-2H]Benzo[a]pyrene, Benzo[a]pyrene D12 |
| InChI Formula | InChI=1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D |
| SMILES | [2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c3c([2H])c([2H])c4c([2H])c([2H])c([2H])c5c([2H])c([2H])c2c3c45 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriobenzo[a]pyrene |
| Molecular Weight (g/mol) | 264.38 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction