missing translation for 'onlineSavingsMsg'
Learn More

Benzo[d]isoxazol-3-ol, 96%, ACROS Organics™

Brand:  Acros Organics 450960050

Additional Details : CAS : 21725-69-9 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15279676

  • 109.10€ / 5g


Estimated Shipment
28-07-2020
Add to basket


Specifications

Chemical Identifiers

21725-69-9
135.12
benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
1,2-benzoxazol-3-one
C7H5NO2
QLDQYRDCPNBPII-UHFFFAOYSA-N
210830
C1=CC=C2C(=C1)C(=O)NO2

Specifications

Benzo[d]isoxazol-3-ol
5g
95% min. (HPLC)
benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
C1=CC=C2C(=C1)C(=O)NO2
135.12
135.12
137°C to 140°C
21725-69-9
C7H5NO2
QLDQYRDCPNBPII-UHFFFAOYSA-N
1,2-benzoxazol-3-one
210830
96%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Benzo¢d!isoxazol-3-ol, 96%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents