missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ NucA Nuclease DuoSet ELISA

Product Code. 30021083
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021083

Brand: R&D Systems™ DY1003805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for the development of sandwich ELISAs to measure natural and recombinant Benzonase Nuclease/NucA

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 23.40 to 1500 pg/mL
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 233874471
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym DcD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.