Learn More
Benzotropine methanesulfonate, Thermo Scientific™
Muscarinic receptor antagonist
Brand:  Thermo Scientific Alfa Aesar J61954.03
 View more versions of this product
Product Code. 15464429
  • ONLINE EXCLUSIVE PRICE
    22.80€
    22.31€ / 1g
    Save 0.49€
    2% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water (81 mg/ml at 25°C), DMSO (50 mg/ml at 25°C), ethanol (81 mg/ml at 25°C), and ether (very slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS Documents