missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog Number p-7058765
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
2495-35-4
C10H10O2
162.19
MFCD00048147
GCTPMLUUWLLESL-UHFFFAOYSA-N
benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
75617
benzyl prop-2-enoate
C=CC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
2495-35-4
C10H10O2
162.19
MFCD00048147
GCTPMLUUWLLESL-UHFFFAOYSA-N
benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
75617
benzyl prop-2-enoate
C=CC(=O)OCC1=CC=CC=C1

Preparation of high refractive index polyacrylates

Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.06 g/mL
Boiling Point 111°C (8 mmHg)
Linear Formula C6H5CH2CO2CH=CH2
Refractive Index 1.518
Quantity 10 g
UN Number UN3082
Solubility Information Miscible with organic solvents and water.
Formula Weight 162.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzyl acrylate, Stabilized with ≈150ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'