missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Benzoate, TRC

Product Code. 17912935
Change view
Click to view available options
Quantity:
1 g
25 g
50 g
Unit Size:
1g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912935 1 g 1g
30137286 50 g 50g
17902395 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17912935 Supplier Toronto Research Chemicals Supplier No. TRCB2309501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-51-4
Formula Weight 212.0837
InChI Formula InChI=1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2
IUPAC Name benzyl benzoate
Molecular Formula C14 H12 O2
Molecular Weight (g/mol) 212.24
Percent Purity >95%
SMILES O=C(OCc1ccccc1)c2ccccc2
Synonym Benzoic acid, benzyl ester (6CI,8CI), Ascabin, Ascabiol, Benylate, Benzyl benzenecarboxylate, Benzyl benzoate, Benzyl phenylformate, Benzylets, Benzyloxy phenyl ketone, Colebenz, NSC 8081, Nicca Sunsolt LM 7EX, Novoscabin, Pelemol B66, Peruscabin, Phenylmethyl benzoate, Scabagen, Scabanca, Scabcare BB, Scabide, Scabiozon, Scobenol, Vanzoate, Venzonate
Chemical Name or Material Benzyl Benzoate
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.