Learn More
Emma Safety Footwear Berlin Safety Shoes
Woven textile combined with PU provides a wear-resistant polyurethane that optimally protects the textile. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot. Thanks to the Hydro-Tec™ lining, this safety shoe is also particularly breathable.
72.70€ - 89.65€
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Gray, Blue |
Design | Berlin |
Liner Material | Hydro-Tec™ Antibacterial |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15948411
|
Emma Safety Footwear
406647-39 |
39 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15958411
|
Emma Safety Footwear
406647-40 |
40 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15968411
|
Emma Safety Footwear
406647-41 |
41 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15978411
|
Emma Safety Footwear
MM40664742 |
42 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15988411
|
Emma Safety Footwear
MM40664743 |
43 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15998411
|
Emma Safety Footwear
MM40664744 |
44 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15908421
|
Emma Safety Footwear
MM40664745 |
45 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15918421
|
Emma Safety Footwear
MM40664746 |
46 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15928421
|
Emma Safety Footwear
MM40664747 |
47 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- S1P-Safety shoe with sole plate for dry work
- Woven Textile Polyurethane coated construction
- Metal-free
- Chrome-free lining
- Hydro-Tec™ Antibacterial lining
- ESD certified
Specifications
EN ISO 20345:2011 | |
Gray, Blue | |
Hydro-Tec™ Antibacterial | |
Composite |
Laces | |
Berlin | |
Textile |