missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | beta 2-Microglobulin |
| Applications | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Formulation | 1x PBS, pH 7.4 |
| Gene Alias | beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Research Discipline | Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?