missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Maltose octaacetate, 98%, Thermo Scientific Chemicals

Catalog Number 11459863
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
22352-19-8
C28H38O19
678.593
MFCD00079432
WOTQVEKSRLZRSX-FKNZMNJBSA-N
Octa-O-acetyl-beta-D-maltose
126963554
[(2S,3R,4S,5R,6S)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
This item is not returnable. View return policy
22352-19-8
C28H38O19
678.593
MFCD00079432
WOTQVEKSRLZRSX-FKNZMNJBSA-N
Octa-O-acetyl-beta-D-maltose
126963554
[(2S,3R,4S,5R,6S)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 157°C to 160°C
Quantity 1 g
Beilstein 79279
Optical Rotation +65° (c=10 in Chloroform)
Formula Weight 678.6
Percent Purity 98%
Chemical Name or Material beta-D-Maltose octaacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'