missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 8 Columns, 3μm Particle Size

Product Code. 10640336 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
50 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10640336

Brand: Thermo Scientific™ 71403154630

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Available in limited dimensions. Please contact Customer Service for more details.

Thermo ScientificTM BetaBasicTM 8 Columns are available in limited dimensions. Please contact Customer Service for more details.

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Ordering Information:

Other column dimensions are also available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
For Use With LC/MS, APCI
Length (Metric) 150 mm
Particle Size 3 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L7
Stationary Phase C8
Product Line Beta
Packing Material Silica
Carbon Load 7%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.