missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 8 Columns, 3μm Particle Size
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
50 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
Description
Thermo ScientificTM BetaBasicTM 8 Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 2.1 mm |
| For Use With | LC/MS, APCI |
| Length (Metric) | 50 mm |
| Particle Size | 3 μm |
| pH | 2 to 9 |
| Pore Size | 150 Å |
| Surface Area | 200 m²/g |
| USP Type | L7 |
| Stationary Phase | C8 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction