Learn More
Saint-Gobain Bev-A-Line V Tubing
Click to view available options
Diameter (Metric) Inner:
1.6 mm
9.6 mm
Diameter (Metric) Outer:
4.8 mm
12.8 mm
Thickness (Metric):
1.6 mm
16.6 mm
Unit Size:
15m
Pack of 15
Description
- Heat-bondable, translucent tubing
- No plasticizers
- Safe for food, beverage, drug, lab and general use
- More affordable than PTFE tubing
- Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350)
- Meets USP Class VI, USDA, and National Formulary requirements
- Sterilize with ethylene oxide or dry heat
- Avoid use with strong solvents
Specifications
Specifications
| Certifications/Compliance | FDA, USP Class VI, USDA, National Formulary requirements |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 9.6 mm |
| Material | Bev-A-Line |
| Diameter (Metric) Outer | 12.8 mm |
| Overall Length (Metric) | 15.2 m |
| Thickness (Metric) | 16.6 mm |
| For Use With (Application) | Safe for food, beverage, drug, lab, and general use |
| Min. Operating Temperature (Metric) | -51°C |
| Packaging Quantity | 1 Each |
| Show More |