missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Big ET-1 ELISA Kit

Product Code. 16392004
Click to view available options
Quantity:
96 Tests
960 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 16392004

Brand: Invitrogen™ BMS2266

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

ELISA

The Human Big ET-1 Platinum ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of human Big ET-1. Cell culture supernatant, serum, and plasma (EDTA, citrate, heparin) have been tested with this assay.

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05305
Assay Range 0.4-25 pg/mL
Assay Sensitivity 0.185 pg/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Vascular Protein
Sample Type Cell Lysates, Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QMEARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME;
Gene ID (Entrez) 1906
Gene Symbol EDN1
Interassay CV 4.9%
Intraassay CV 4.7%
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, Streptavidin-HRP, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases EDN1; endothelin 1; Endothelin-1; ET1; HDLCQ7; PPET1; preproendothelin-1
Quantity 96 Tests
Regulatory Status RUO
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target Endothelin 1
Test Time 1 hr 20 min
Total Assay Time 4 hr
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.