Learn More
Zeiss™ Binocular Ergo Phototube
Product Code. 11845113
Click to view available options
Type:
Binocular Ergophototube, 20/23° (100:0/0:100)
Unit Size:
Each
Product Code. 11845113
Brand: Zeiss™ 4255120000000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Features reversed image variable and continuous vertical adjustment up to 44mm.
Compatible Microscopes:
  • Axio Imager.A1
  • Axio Imager.A1m
  • Axio Imager.A2
  • Axio Imager.D1
  • Axio Imager.D1 for epi-fluorescence with ApoTome equipment
  • Axio Imager.D1m
  • Axio Imager.D2
  • Axio Imager.M1
  • Axio Imager.M1m
  • Axio Imager.M2
  • Axio Imager.Z1
  • Axio Imager.Z1 + ApoTome
  • Axio Imager.Z1m
  • Axio Imager.Z2
  • Axio Lab.A1
  • Axio Lab.A1 FL-LED
  • Axio Lab.A1 MAT
  • Axio Lab.A1 Pol
  • Axio Scope.A1
  • Axio Scope.A1 Pol
  • Axio Scope.A1 Vario
Specifications
Type Binocular Ergophototube, 20/23° (100:0/0:100)
Includes Camera port with interface 60N
Quantity 1 Ea.