Learn More
Zeiss™ Binocular Phototube
Product Code. 11814983
Click to view available options
Type:
Binocular Phototube 20°/23 (100:0/0:100)
Binocular Phototube 30°/20 (50:50)
Binocular Phototube 30°/23 (100:0/0:100)
Binocular Phototube 30°/23 (50:50)
Binocular Phototube S 20°
Binocular Phototube, Left Port 45°/23 (50:50)
Binocular Tube 20°/23
Binocular Tube 30°/20
Unit Size:
Each
Product Code. 11814983
Brand: Zeiss™ 4255209030000
Use these phototubes for a variety of applications
Binocular phototubes are available with a variety of options to suit your needs
Specifications
Type Binocular Phototube 20°/23 (100:0/0:100)
Description Upright image with sliding prism, camera port with interface 60N
Quantity 1 Ea.