Learn More
Zeiss™ Binocular Phototube
Click to view available options
Type:
Binocular Phototube 20°/23 (100:0/0:100)
Binocular Phototube 30°/23 (100:0/0:100)
Binocular Phototube S 20°
Binocular Phototube, Left Port 45°/23 (50:50)
Binocular Tube 20°/23
Binocular Tube 30°/20
Unit Size:
Each
Description
Binocular phototubes are available with a variety of options to suit your needs
Specifications
Specifications
| Type | Binocular Tube 20°/23 |
| Description | Upright image |
| Quantity | 1 Ea. |