Learn More
Zeiss™ Binocular Phototube
Click to view available options
Type:
Binocular Phototube 20°/23 (100:0/0:100)
Binocular Phototube 30°/23 (100:0/0:100)
Binocular Phototube S 20°
Binocular Phototube, Left Port 45°/23 (50:50)
Binocular Tube 20°/23
Binocular Tube 30°/20
Unit Size:
Each
Description
Binocular phototubes are available with a variety of options to suit your needs
Specifications
Specifications
| Type | Binocular Phototube S 20° |
| Description | For 51 to 75mm eye distance |
| Includes | With 100/100 switchover of the right channel to interface 60N |
| Quantity | 1 Ea. |