missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Indigo™ Sterile Nitrile Gloves

Catalog Number 15278208
Click to view available options
Size:
10
5, 5.5
6, 6.5
7, 7.5
8, 8.5
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Feature textured fingers for enhanced grip. Nitritex™ BioClean Indigo™ Sterile Nitrile Gloves are latex free gloves that are processed to ensure ISO Class 4 compatibility, are resistant to a wide range of chemicals and offer excellent antistatic properties.

  • Resistant to a wide range of chemicals
  • Textured fingers for enhanced grip
  • Powder-free and latex-free
  • Excellent antistatic properties
  • Beaded cuff
  • Non-particulating EasyTear™ packaging
Certifications/Compliance (Europe) ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, ISO 11137-1:2006, (EC)1935/2004 and (EC)2023/2006, ASTM D6978-05
Cleanroom Class Class 10 (ISO 4)
Color Purple
Disposable Single-use
Exterior Finish Fingertip Textured
Grade EU GMP Grade A
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 6, 6.5
Sterility Sterile
Glove Cuff Style Beaded
Model BNPLS6065
Packaging Type EasyTear™
Quantity One pair per inner PE wallet; one wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs)
Resistant To Chemicals
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'