Learn More
Molecular Probes™ Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Brand:  Molecular Probes™ T12921
Additional Details : Weight : 0.00450kg
Product Code. 11520756
  • 438.00€ / 5mg
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
The water-soluble neuronal tracer 5-(and-6)-tetramethylrhodamine biocytin (biocytin TMR) can be detected in fixed tissues by using avidin or streptavidin probes. However, tetramethylrhodamine biocytin can also be observed directly in fixed or unfixed samples because the attached tetramethylrhodamine dye produces bright, orange-fluorescence (absorption/emission maxima ∼554/581 nm).
Specifications
Specifications
Classic Dyes
Room Temperature
Invitrogen™
Store at room temperature and protect from light.
Solid
Biocytins
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.