missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation BioResolve RP mAb Polyphenyl Method Validation Kit
Click to view available options
Length (Metric):
100 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Pack of 3
Description
- The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
- This Method Validation Kit Part Number contains (3) columns from (3) different manufactured batches.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 150 mm |
| Particle Size | 2.7 μm |
| Pore Size | 450 Å |
| Quantity | 3/Pk. |
| Type | Method Validation Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction