Learn More
IBA Lifesciences Biotin Blocking Buffer
Product Code. 12733064 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
2 mL
Unit Size:
Each
Product Code. 12733064
Brand: IBA Lifesciences 20501002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Biotin Blocking Buffer is optimized for efficient blocking of biotinylated proteins in western blot analyses
Depending on the host organism, protein extracts may contain small amounts of biotinylated proteins such as the biotin carboxyl carrier protein (BCCP, 22 kDa) of E. coli.Biotin Blocking Buffer is recommended for blocking such biotinylated proteins on western blots prior to detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins with Strep-Tactin™ or Strep-Tactin™XT conjugates to prevent the detection of false positive bands. The buffer contains the tetrameric biotin binding protein avidin, which selectively binds biotinylated proteins. Please note that usage of BioLock is not necessary when using StrepMAB-Classic, since this antibody does not bind to biotinylated proteins.
Specifications
Quantity 2 mL