missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Biotinylated Recombinant Human IL-7 Protein

Product Code. 18755935
Change view
Click to view available options
:
25 μg
Product Code.
18755935 25 μg
1 options
This item is not returnable. View return policy

Product Code. 18755935

Brand: R&D Systems™ BTN007025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human IL-7 (Catalog # BTN007) binds to Recombinant Human IL-7R alpha/CD127 Fc Chimera (Catalog # 10531-IR ) with a ED50 of 0.200-2.00 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13232.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) MolecularWeight-observed: 15-28 kDa, under reducing conditions., MolecularWeight-theroretical: 17 kDa
Quantity 25 μg
Source E. coli-derived human IL-7 protein Asp26-His177
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL7, IL-7 interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Conjugate Biotin
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.