Learn More
Bis(2-dimethylaminoethyl) ether, 98%
Product Code. 11372327
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11372327
Brand: Thermo Scientific Alfa Aesar L09884.09
Request Bulk or Custom Format
CAS: 3033-62-3 | C8H20N2O | 160.26 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3033-62-3
Molecular Formula C8H20N2O
Molecular Weight (g/mol) 160.26
MDL Number MFCD00059199
InChI Key GTEXIOINCJRBIO-UHFFFAOYSA-N
Synonym bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
PubChem CID 18204
IUPAC Name 2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOCCN(C)C
Specifications
Boiling Point 179°C to 180°C
Flash Point 66°C (151°F)
Quantity 10 g
UN Number UN2922
Beilstein 1739668
Formula Weight 160.26
Percent Purity 98%
Chemical Name or Material Bis(2-dimethylaminoethyl) ether
compliance-icons compliance-icons
Product Identifier
  • Bis(2-dimethylaminoethyl) ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Bis(2-dimethylaminoethyl) ether