missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-dimethylaminoethyl) Ether, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2,2'-Oxybis(N,N-dimethylethanamine) |
| CAS | 3033-62-3 |
| Molecular Formula | C8 H20 N2 O |
| Quantity | 10 g |
| Synonym | Ethanamine, 2,2'-oxybis[N,N-dimethyl-, Ethylamine, 2,2'-oxybis[N,N-dimethyl- (6CI,7CI,8CI), 2,2'-Oxybis[N,N-dimethylethanamine], 1,5-Bis(dimethylamino)-3-oxapentane, 2,2'-Oxybis[N,N-dimethylethylamine], A 1, A 133, A 99, A 99 (amine), Addocat 108, BL 11, BL 19, Bis(dimethylaminoethyl) ether, Bis[2-(dimethylamino)ethyl] ether, DY 1, Dabco BL 11, Dabco BL 19, Dabco BL 19I, ET 33B, Jeffcat Z 20, Jeffcat ZF 20, Jeffcat ZM 20, Kalpur PC, Kaolizer 12, Kaolizer 12P, Minico TMDA, N,N,N',N'-Tetramethyl-2,2'-oxybis[ethylamine], N,N,N',N'-Tetramethyl-3-oxapentane-1,5-diamine, N,N,N',N'-Tetramethylbisaminoethyl ether, NP 99, NSC 109887, Niax A 1, Niax A 99, Niax Catalyst A 1, PC CAT NP 133, PC CAT NP 90, PC CAT NP 99, Tetramethyl bis(aminoethyl) ether, Texacat ZF 20, Texacat ZF 22, Toyocat ET, Toyocat ET 33B, Toyocat ETS, ZF 20, 2,2'-Oxybis(N,N-dimethylethanamine) |
| InChI Formula | InChI=1S/C8H20N2O/c1-9(2)5-7-11-8-6-10(3)4/h5-8H2,1-4H3 |
| SMILES | CN(C)CCOCCN(C)C |
| IUPAC Name | 2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine |
| Molecular Weight (g/mol) | 160.257 |
| Formula Weight | 160.1576 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction