missing translation for 'onlineSavingsMsg'
Learn More

Bis(dicarbonylcyclopentadienyliron), 99%, Thermo Scientific Chemicals

Catalog Number p-7049908
Click to view available options
Quantity:
5 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12154-95-9
MFCD00135625
Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12154-95-9
MFCD00135625
Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl

Cyclopentadienyliron dicarbonyl dimer is a workhorse in organometallic chemistry because it is inexpensive and rugged. It is also used as a pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Melting Point 194°C (decomposition)
Color Red
Quantity 5 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information It is readily soluble in moderate polar organic solvents such as chloroform and pyridine,but less soluble in carbon tetrachloride and carbon disulfide. It is sparingly soluble in water.
Formula Weight 353.93
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)
compliance-icons
Product Identifier
  • Bis(dicarbonylcyclopentadienyliron)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'