Learn More
bis-Tris, ≥99%, MP Biomedicals™
Catalog Number p-4403876
166.15 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
6976-37-0
C8H19NO5
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
6976-37-0
C8H19NO5
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
Exhibits a very small shift in dissociation constant with respect to temperature
  • Zwitterionic buffer
  • Buffering agent
  • White Powder
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range >99%
Synonym 2,2-bis(Hydroxymethyl)-2,2',2”-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2'
Formula Weight 209.2
Chemical Name or Material bis-Tris
compliance-icons
Product Identifier
  • Bis-Tris
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..