missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Carbonate Basic, purum p.a., 80 to 82% Bi Basis (KT), Honeywell™

Product Code. p-7120226 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15667550

Brand: Honeywell Research Chemicals 95376250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
MDL Number MFCD00010877
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O

Specifications

Quantity 250 g
Linear Formula (BiO)2CO3
UN Number NONH for all modes of transport
Formula Weight 509.97g/mol
Percent Purity 80 to 82% (Bi basis) (KT)
Packaging Poly Bottle
Density 6.86g/mL at 25°C
Chemical Name or Material Bismuth(III) carbonate basic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.