missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ BiTek™ Fermentation Broth
Description
Gibco™ BiTek™ Fermentation Broth is a bacterial fermentation formulated with ingredients that exhibit superior consistency, performance, and solubility.
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Bacterial fermentation |
| Product Line | BiTek |
| Quantity | 10 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?