Learn More
Biuret Reagent, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Quantity | 100 mL |
Safety and Handling
Product Identifier
- BIURET REAGENT
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : copper sulphate, lead di(acetate)