missing translation for 'onlineSavingsMsg'
Learn More

Black Eriochrome T, Chem Lab™

Product Code. 11993323
Click to view available options
Concentration:
10 g/L
5 g/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11993323

Brand: Chem-Lab CL06.0501.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Concentration 10 g/L
Solubility Information Soluble in water
Odor Ethanolic
Color Purple
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Eriochrome black T indicator
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.