Learn More
Kautex™ Black Polypropylene Screw Closures
Click to view available options
For Use With (Equipment):
100 mL Bottle
1500/2500/4000 mL Bottle
200/300 mL Bottle
2000 mL Bottle
250/350 mL Bottle
50 mL Bottle
500 mL Bottle
500/750/1000 mL Bottle
750/1000 mL Bottle
Diameter (Metric):
25 mm
32 mm
40 mm
45 mm
50 mm
54 mm
65 mm
80 mm
Includes:
w/Tamper Evident Ring
w/out Tamper Evident Ring
Unit Size:
Each
Pack of 100
Spécification
Spécification
| Color | Black |
| Includes | w/Tamper Evident Ring |
| Material | Polypropylene |
| Thread Size (Metric) | 00 mm |
| Liner Material | Polyethylene Foam Insert |
| Neck Finish | 00 mm |
| Product Type | Screw Cap |
| For Use With (Equipment) | 100 mL Bottle |
| Diameter (Metric) | 40 mm |