Learn More
Pacific Handy Cutter Blade Disposal with Wall Bracket
Pacific Handy Cutter Blade Bank is the safest and most convenient way to correctly dispose of used razor blades. When utilized throughout the workplace the Blade Banks eliminate the potential liabilities and safety risks caused by loose blades.
Brand: Pacific Handy Cutter BH-00206
Additional Details : Weight : 0.01430kg
Description
- This key component of our Safety First System keeps blades out the reach of customers, off the ground, out of trash bags and all other potentially hazardous locations.
- Safely contain over 750 used blades and can be easily mounted to walls, posts, pallet racks and more.
- By pairing one of PHC's Blade Dispensers (SPD-017, QBD-324, or QBD-379) to the front of the Blade Bank, you create a convenient blade change station that will increase the safety and efficiency of your staff.
- By using both items together, your employees will be encouraged to properly dispose of loose blades and easily access new replacement blades near their work stations and throughout your facility, ultimately increasing overall productivity.
- The BH-00206 includes the wall mount bracket and mounting screws.
Specifications
Sharps Container | |
0.8 kg | |
Not Autoclavable |
Plastic | |
Yellow |