Learn More
Brand™ Silberbrand™ Borosilicate Glass Class B Volumetric Flask
Product Code. 10291831 Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
Closure Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Limit of Error:
±0.06 mL
±0.09 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
±0.9 mL
±1.8 mL
Unit Size:
Each
Pack of 2
Product Code. 10291831
Brand: Brand™ 36754
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Manufactured from Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Silberbrand™ Borosilicate Glass Class B Volumetric Flask includes polypropylene stopper. White enamel DE-M marking provides quick volume check.
  • Manufactured from Borosilicate Glass
  • Complies with DIN EN ISO 1042
  • Class B
  • Calibrated to contain (TC, In.)
  • Includes polypropylene stopper
  • White enamel DE-M marking provides quick volume check
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042
Closure Material Polypropylene
Color Clear
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Limit of Error ±0.9 mL
No. per Case 1 Piece
Tolerance ±0.9 mL
Class B
Closure Size 29/32