Learn More
BRAND™ Blaubrand™ Amber Borosilicate Glass Class A Volumetric Flask, with Certificate
Product Code. 15825552 Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 15825552
Brand: BRAND™ 937407
Only null left
Add to basket
Add to basket
Manufactured from Amber Borosilicate Glass and complies with DIN EN ISO 1042 standards. Includes a polypropylene stopper, but is interchangeable with amber glass stopper. The dark flask protects against deterioration of light-sensitive liquids. Delivered with individual conformity certificate.
  • Manufactured from Amber Borosilicate Glass.
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • The dark glass protects against deterioration of light-sensitive liquids.
  • Includes polypropylene stopper, but is interchangeable with amber glass stopper.
  • Batch number and certificate included. Delivered with individual conformity certificate.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042. DAkkS calibration and USP certificates upon request.
Closure Material Polypropylene
Color Amber
Capacity (Metric) 100 mL
Material Borosilicate Glass
Limit of Error ±0.10 mL
Neck Style Narrow
No. per Case 2 Piece
Tolerance ±0.10 mL
Class A
Closure Size 14/23