Learn More
Macherey-Nagel™ Blotting paper MN 218 B
Product Code. p-8013702
Click to view available options
Dimensions (L x W):
150 x 200 mm
200 x 200 mm
Unit Size:
Pack of 100
Product Code. 11992292
Brand: Macherey-Nagel™ 742138
Market leading paper quality for reliable biomolecule blottings – MN 218 B
  • Speed: Slow
Specifications
Type Blotting Paper
Quantity 100 Sheets
Thickness (Metric) 0.36 mm
Dimensions (L x W) 150 x 200 mm