Learn More
ABEBA™ Blue Autoclavable Clogs 9610
Antistatic and autoclavable at 121°C. High slip resistance.
Brand:  ABEBA™ 9610-45/46
5.80 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15106956
  • 50.95€ / 1 pair
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Blue
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRC
9610
Controlled Environments Shoes
Blue
Static