Learn More
Fisherbrand™ Blue Micro PTFE Stir Bar
Product Code. 16245841 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
6 mm
Unit Size:
Pack of 10
Product Code. 16245841
Only null left
Add to basket
Add to basket
Fisherbrand™ Blue Micro PTFE Stir Bar is an inert and leak-free stirrer for a wide variety of applications where identification is important.
  • Micro stirrer bars
  • For use where identification is of prime importance
  • PTFE coatings are not as inert as pure PTFE
  • Individually polished to stay cleaner for longer
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Blue
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 3 mm
Length (Metric) 6 mm
Quantity 10/Pk.
Shape Micro