Learn More
3M™ Blue with White Back Panel SMS Based Polypropylene Protective Coveralls
120.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Size:
Large
X-Large
Specifications
Product Type | Protective Coveralls |
Size | Large |
Waist Style | Elastic |
Neck Style | Hooded |
Hood Type | 3-panel |
Protection Class | Category III |
Resistance | Flame |
Height (Metric) | 174 to 181 cm |
Style | Enhanced Gusset |