missing translation for 'onlineSavingsMsg'
Learn More

BNP Antibody (50E1cc), DyLight 755, Novus Biologicals™

Product Code. 18210392 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18210392 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18210392 Supplier Novus Biologicals Supplier No. NB200440IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 50E1cc
Conjugate DyLight 755
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Host Species Mouse
Immunogen Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4879
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.