missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | BNP |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 50E1cc |
| Conjugate | Janelia Fluor 549 |
| Dilution | ELISA, Sandwich ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein |
| Host Species | Mouse |
| Immunogen | Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?