Learn More
Helios Italquartz Boats with Ring Handle, Transparent
Product Code. 15848034
Click to view available options
Dimensions (L x W):
120 mm x 15 mm
143 mm x 17 mm
180 mm x 15 mm
76 mm x 10 mm
76 mm x 11 mm
Unit Size:
Each
Product Code. 15848034
Brand: Helios Italquartz 62P00004
Only null left
Add to basket
Add to basket
Boats with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
  • Transparent
  • Include ring handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Bottom Shape Flat
Color Transparent
Material Quartz
Shape Boat
Type Weighing Boat with Ring Handle
Dimensions (L x W) 76 mm x 10 mm
Length (Metric) 76/16 mm
Width (Metric) 10 mm
Height (Metric) 10 mm