Learn More
Helios Italquartz Boats with Ring Handle, Transparent
Click to view available options
Dimensions (L x W):
120 mm x 15 mm
143 mm x 17 mm
180 mm x 15 mm
76 mm x 10 mm
76 mm x 11 mm
Unit Size:
Each
Description
- Transparent
- Include ring handle
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Specifications
| Bottom Shape | Flat |
| Color | Transparent |
| Material | Quartz |
| Shape | Boat |
| Type | Weighing Boat with Ring Handle |
| Dimensions (L x W) | 76 mm x 11 mm |
| Length (Metric) | 76/19 mm |
| Width (Metric) | 11 mm |
| Height (Metric) | 11 mm |