Learn More
Bohlender™ BOLA™ Joint Clamps
Product Code. 17197414
Click to view available options
Size:
NS 45/50
Unit Size:
Pack of 3
Product Code. 17197414
Brand: Bohlender™ H94245
Only null left
Add to basket
Add to basket
Joint Clamps with steel spring core for connection of standard ground joints.
  • PTFE encapsulated steel spring core
  • Universal chemical resistance since product is only exposed to PTFE
Specifications
Material PTFE
For Use With (Application) Connection of Standard Ground Joints
Size NS 45/50