Learn More
Thermo Scientific™ Bonded AVCS Pre-slit Caps
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
Brand:  Thermo Scientific™ C5000-72B
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 15378026
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
  • Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases.
  • AVCS closures seal as well as crimp top vials.
  • AVCS design allows for consistent sealing, eliminating user-to-user variance.
Specifications
Specifications
Blue
Yes
Surestop
9 mm
PTFE/Silicone
100/Pk.