Learn More
Thermo Scientific™ Bonded AVCS Pre-slit Caps
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
38.93€ - 45.63€
Specifications
Diameter (Metric) Outer | 9 mm |
---|---|
Pre-slit | Yes |
Septum | PTFE/Silicone |
Quantity | 100/Pk. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
15328036
|
Thermo Scientific™
C5000-72Y |
Yellow |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15318036
|
Thermo Scientific™
C5000-72R |
Red |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15398026
|
Thermo Scientific™
C5000-72G |
Green |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15378026
|
Thermo Scientific™
C5000-72B |
Blue |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
- Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases.
- AVCS closures seal as well as crimp top vials.
- AVCS design allows for consistent sealing, eliminating user-to-user variance.
Specifications
9 mm | |
Yes | |
PTFE/Silicone | |
100/Pk. |