missing translation for 'onlineSavingsMsg'
Learn More

Boron powder, 98% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 040036.03

Quantity 1g

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15426315

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7440-42-8
10.81
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
5462311
boron
B
MFCD00134034
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
CHEBI:27560
[B]

Specifications

7440-42-8
98% (Metals basis)
MFCD00134034
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron
5462311
Odorless
Boron
1g
B
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
[B]
10.81
CHEBI:27560
Brown powder
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.