Learn More
Laboratory Sales™ Borosilicate Glass Bottle Crimp Vial
Shop All Wheaton Science Products Products
Click to view available options
Unit Size:
466 sets
Description
- Ideal for securely containing contents with solid or septa caps.
- Transparent for content visibility.
Specifications
Specifications
| Diameter (Metric) Outer | 15 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 2 mL |
| No. per Pack | 466 |
| Height (Metric) | 33 mm |
| Quantity | 466 Pk. |