Learn More
Rogo Sampaic™ Glass Coverslips
Product Code. 13206718
Click to view available options
Length (Metric):
12 mm
14 mm
18 mm
22 mm
24 mm
Material:
Borosilicate Glass
Soda Lime Glass
Quantity:
100/Pk.
200/Pk.
Unit Size:
Pack of 100
Pack of 200
Product Code. 13206718
Brand: Rogo Sampaic™ BPD006
Only null left
Add to basket
Add to basket
Preserve and protect specimens on slides.
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Coverslip
Shape Rectangle and Square
Material Soda Lime Glass
Length (Metric) 24 mm
Quantity 100/Pk.